Learn More
Biomatik Corporation Mouse C-type lectin domain family 18 member A(CLEC18A) ELISA kit
Supplier:  Biomatik Corporation EKC36680
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q7TSQ1. Target Synonyms: FLJ77955, MGC164804, MRCL, MRCL1, mannose receptor-like 1|secretory protein LOC348174. Restriction: For Research Use Only.
Catalog No. 50-229-4392