Learn More
Biomatik Corporation Mouse C-X-C Motif Chemokine 11 (Cxcl11) Elisa Kit
Supplier:  Biomatik Corporation EKC36687
Encompass_Preferred
Mouse C-X-C Motif Chemokine 11 (Cxcl11) Elisa Kit for detecting Chemokine (C-X-C Motif) Ligand 11 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9JHH5; Cxcl11. Target Synonyms: Cxcl11; Scyb11C-X-C motif chemokine 11; Interferon-inducible T-cell alpha chemoattractant; I-TAC; Small-inducible cytokine B11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4394