Learn More
Biomatik Corporation Mouse Cytochrome-C, Cyt-C Elisa Kit
Supplier:  Biomatik Corporation EKC36707
Encompass_Preferred
Mouse Cytochrome-C, Cyt-C Elisa Kit for detecting Cytochrome C, Somatic in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 34.3-2200pg/ml. Sensitivity: 8.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P62897; Cycs. Target Synonyms: Cycs; Cytochrome c; somatic. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4396