Learn More
Biomatik Corporation Mouse Ectonucleotide pyrophosphatase/phosphodiesterase family member 2(ENPP2) ELISA kit
Supplier:  Biomatik Corporation EKC36779
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 72 ng/mL-2500 ng/mL. Sensitivity: 36 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9R1E6. Target Synonyms: ATX, ATX-X, AUTOTAXIN, FLJ26803, LysoPLD, NPP2, PD-IALPHA, PDNP2, autotaxin|autotaxin-t|phosphodiesterase I/nucleotide pyrophosphatase 2|plasma lysophospholipase D. Restriction: For Research Use Only.
Catalog No. 50-229-4402