Learn More
Biomatik Corporation Mouse Ezrin(EZR) ELISA kit
Supplier:  Biomatik Corporation EKC36837
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 62.5 pg/mL-4000 pg/mL. Sensitivity: 15.6 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P26040. Target Synonyms: CVIL, CVL, DKFZp762H157, FLJ26216, MGC1584, VIL2, cytovillin 2|villin 2 (ezrin). Restriction: For Research Use Only.
Catalog No. 50-229-4404