Learn More
Biomatik Corporation Mouse Follistatin, Fs Elisa Kit
Supplier:  Biomatik Corporation EKC36895
Encompass_Preferred
Mouse Follistatin, Fs Elisa Kit for detecting Follistatin in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.05-10ng/ml. Sensitivity: 0.025ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P47931; Fst. Target Synonyms: FstFollistatin; FS; Activin-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4410