Learn More
Biomatik Corporation Mouse Granulocyte-Macrophage Colony Stimulating Factor,GM-CSF ELISA Kit
Supplier:  Biomatik Corporation EKC36990
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P01587. Target Synonyms: GMCSF, MGC131935, MGC138897, colony stimulating factor 2|granulocyte-macrophage colony stimulating factor|molgramostin|sargramostim. Restriction: For Research Use Only.
Catalog No. 50-229-4416