Learn More
Biomatik Corporation Mouse Lymphocyte activation gene 3 protein(LAG3) ELISA kit
Supplier:  Biomatik Corporation EKC37313
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 1.25 ng/mL-80 ng/mL. Sensitivity: 0.31 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q61790. Target Synonyms: CD223, lymphocyte-activation protein 3. Restriction: For Research Use Only.
Catalog No. 50-229-4428