Learn More
Biomatik Corporation Mouse Macrophage Mannose Receptor 1 (Mrc1) Elisa Kit
Supplier:  Biomatik Corporation EKC37336
Encompass_Preferred
Mouse Macrophage Mannose Receptor 1 (Mrc1) Elisa Kit for detecting Mannose Receptor, C Type 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q61830; Mrc1. Target Synonyms: Mrc1; Macrophage mannose receptor 1; MMR; CD antigen CD206. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4433