Learn More
Biomatik Corporation Mouse Matrix Metalloproteinase 8/Neutrophil Collagenase, Mmp-8 Elisa Kit
Supplier:  Biomatik Corporation EKC37362
Encompass_Preferred
Mouse Matrix Metalloproteinase 8/Neutrophil Collagenase, Mmp-8 Elisa Kit for detecting Matrix Metallopeptidase 8 (Neutrophil Collagenase) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O70138; Mmp8. Target Synonyms: Mmp8; Neutrophil collagenase; EC 3.4.24.34; Collagenase 2; Matrix metalloproteinase-8; MMP-8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4438