Learn More
Biomatik Corporation Pig Creatine kinase M-type(CKM) ELISA kit
Supplier:  Biomatik Corporation EKC42433
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Sus scrofa (Pig). Detection Range: 0.156 mU/mL-10 mU/mL. Sensitivity: 0.039 mU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q5XLD3. Target Synonyms: CKMM, M-CK, creatine kinase M chain|creatine kinase-M|muscle creatine kinase. Restriction: For Research Use Only.
Catalog No. 50-229-5079