Learn More
Biomatik Corporation Sheep Corticoliberin(CRH) ELISA kit
Supplier:  Biomatik Corporation EKC42445
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Ovis aries (Sheep). Detection Range: 14 pg/mL-500 pg/mL. Sensitivity: 14 pg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P01142. Target Synonyms: CRF, corticotropin-releasing factor. Restriction: For Research Use Only.
Catalog No. 50-229-5082