Learn More
Biomatik Corporation Human Receptor Tyrosine-Protein Kinase Erbb-3 (Erbb3) Elisa Kit
Supplier:  Biomatik Corporation EKC42470
Encompass_Preferred
Human Receptor Tyrosine-Protein Kinase Erbb-3 (Erbb3) Elisa Kit for detecting V-Erb-B2 Erythroblastic Leukemia Viral Oncogene Homolog 3 (Avian) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P21860; ERBB3. Target Synonyms: c erbB 3; c erbB3; Erb b2 receptor tyrosine kinase 3; ErbB 3; ERBB3; ERBB3 protein; erbB3 S; ERBB3_HUMAN; Glial growth factor receptor; HER 3; HER3; Human epidermal growth factor receptor 3; LCCS2; MDA BF 1; MGC88033; p180 ErbB3; p45 sErbB3; p85 sErbB3; proto-oncogene-like protein c ErbB 3; proto-oncogene-like protein c ErbB3; Proto-oncogene-like protein c-ErbB-3; Receptor tyrosine protein kinase erbB 3; Receptor tyrosine protein kinase erbB3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5091