Learn More
Biomatik Corporation Rat Follistatin-related protein 1(FSTL1) ELISA kit
Supplier:  Biomatik Corporation EKC42487
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.6 ng/mL-1000 ng/mL. Sensitivity: 3.9 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q62632. Target Synonyms: FLJ50214, FLJ52277, FRP, FSL1, follistatin-related protein. Restriction: For Research Use Only.
Catalog No. 50-229-5098