Learn More
Biomatik Corporation Human Interleukin enhancer-binding factor 3(ILF3) ELISA kit
Supplier:  Biomatik Corporation EKC42532
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates, cell lysates. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q12906. Target Synonyms: CBTF, DRBF, DRBP76, MMP4, MPHOSPH4, MPP4, NF-AT-90, NF110, NF90, NFAR, NFAR-1, NFAR2, TCP110, TCP80, M-phase phosphoprotein 4|double-stranded RNA-binding protein, 76 kD|dsRNA binding protein NFAR-2/. Restriction: For Research Use Only.
Catalog No. 50-229-5111