Learn More
Biomatik Corporation Rat Lysozyme C(LYZ) ELISA kit
Supplier:  Biomatik Corporation EKC42561
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P61626. Target Synonyms: LZM, lysozyme, 1,4-beta-N-acetylmuramidase C|lysozyme|lysozyme C. Restriction: For Research Use Only.
Catalog No. 50-229-5122