Learn More
Biomatik Corporation Mouse Midkine(MDK) ELISA kit
Supplier:  Biomatik Corporation EKC42569
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.81 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P12025. Target Synonyms: FLJ27379, MK, NEGF2, OTTHUMP00000197041|OTTHUMP00000197042|OTTHUMP00000197045|midkine. Restriction: For Research Use Only.
Catalog No. 50-229-5124