Learn More
Biomatik Corporation Human Matrix Metalloproteinase-28 (Mmp28) Elisa Kit
Supplier:  Biomatik Corporation EKC42574
Encompass_Preferred
Human Matrix Metalloproteinase-28 (Mmp28) Elisa Kit for detecting Matrix Metallopeptidase 28 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9H239; MMP28. Target Synonyms: Epilysin; Matrix metalloproteinase 28; Matrix metalloproteinase-28; MM 28; MM28; MMP 25; MMP 28; MMP-28; MMP25; MMP28; MMP28_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5128