Learn More
Biomatik Corporation Human Neogenin (Neo1) Elisa Kit
Supplier:  Biomatik Corporation EKC42583
Encompass_Preferred
Human Neogenin (Neo1) Elisa Kit for detecting Neogenin Homolog 1 (Chicken) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q92859; NEO1. Target Synonyms: Immunoglobulin superfamily DCC subclass member 2; NEO1; NEO1_HUMAN; Neogenin; Ngn (gene name). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5131