Learn More
Biomatik Corporation Mouse neuregulin-4 (NRG4) ELISA kit
Supplier:  Biomatik Corporation EKC42589
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 3.12 ng/mL-200 ng/mL. Sensitivity: 0.78 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9WTX4. Target Synonyms: DKFZp779N0541, DKFZp779N1944, HRG4, heregulin 4. Restriction: For Research Use Only.
Catalog No. 50-229-5133