Learn More
Biomatik Corporation Fish Trypsin Elisa Kit
Supplier:  Biomatik Corporation EKC42601
Encompass_Preferred
Fish Trypsin Elisa Kit for detecting Protease, Serine, 1 (Trypsin 1) in serum, plasma, tissue homogenates. Species Reactivity: Fish (Vertebrata). Detection Range: 0.312-20ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P35031. Target Synonyms: Trypsin-1; EC 3.4.21.4; Trypsin I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5137