Learn More
Biomatik Corporation Mouse serum amyloid A2 (SAA2) ELISA kit
Supplier:  Biomatik Corporation EKC42610
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 7.8 ng/mL-500 ng/mL. Sensitivity: 1.95 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P05367. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-5140