Learn More
Biomatik Corporation Human Serpin B5 (Serpinb5) Elisa Kit
Supplier:  Biomatik Corporation EKC42627
Encompass_Preferred
Human Serpin B5 (Serpinb5) Elisa Kit for detecting Serpin Peptidase Inhibitor, Clade B (Ovalbumin), Member 5 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P36952; SERPINB5. Target Synonyms: Maspin; Ovalbumin; Peptidase inhibitor 5; PI-5; PI5; protease inhibitor 5 (maspin); Protease inhibitor 5; Serine (or cysteine) peptidase inhibitor; clade B; member 5; serine (or cysteine) proteinase inhibitor; clade B (ovalbumin); member 5; Serpin B5; serpin family B member 5; Serpin peptidase inhibitor; Serpin peptidase inhibitor; clade B (ovalbumin); member 5; Serpinb5; SPB5_HUMAN; Spi5; Spi7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5144