Learn More
Biomatik Corporation Mouse Secreted Frizzled-Related Protein 4 (Sfrp4) Elisa Kit
Supplier:  Biomatik Corporation EKC42631
Encompass_Preferred
Mouse Secreted Frizzled-Related Protein 4 (Sfrp4) Elisa Kit for detecting Secreted Frizzled-Related Protein 4 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.47-30ng/ml. Sensitivity: 0.11ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9Z1N6; Sfrp4. Target Synonyms: Sfrp4; Secreted frizzled-related sequence protein 4; FRP-4; sFRP-4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5146