Learn More
Biomatik Corporation Human Sialic Acid-Binding Ig-Like Lectin 8 (Siglec8) Elisa Kit
Supplier:  Biomatik Corporation EKC42636
Encompass_Preferred
Human Sialic Acid-Binding Ig-Like Lectin 8 (Siglec8) Elisa Kit for detecting Sialic Acid Binding Ig-Like Lectin 8 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9NYZ4; SIGLEC8. Target Synonyms: CD329 antigen; CDw329; SAF 2; SAF-2; SAF2; Sialic acid binding Ig like lectin 8; Sialic acid-binding Ig-like lectin 8; Sialoadhesin family member 2; SIGL8_HUMAN; Siglec 8; Siglec-8; SIGLEC8; SIGLEC8L. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5148