Learn More
Biomatik Corporation Human Kunitz-Type Protease Inhibitor 1 (Spint1) Elisa Kit
Supplier:  Biomatik Corporation EKC42649
Encompass_Preferred
Human Kunitz-Type Protease Inhibitor 1 (Spint1) Elisa Kit for detecting Serine Peptidase Inhibitor, Kunitz Type 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O43278; SPINT1. Target Synonyms: HAI 1; HAI; HAI-1; HAI1; Hepatocyte growth factor activator inhibitor type 1; Kunitz type protease inhibitor 1; Kunitz-type protease inhibitor 1; MANSC2; Serine peptidase inhibitor Kunitz type 1; SPINT1; SPIT1_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Numéro de catalogue. 50-229-5157
Ajouter au panier
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.