Learn More
Biomatik Corporation Mouse Stanniocalcin-1 (Stc1) Elisa Kit
Supplier:  Biomatik Corporation EKC42654
Encompass_Preferred
Mouse Stanniocalcin-1 (Stc1) Elisa Kit for detecting Stanniocalcin 1 in serum, plasma, tissue homogenates, cell culture supernates. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O55183; Stc1. Target Synonyms: Stc1; Stc; Stanniocalcin-1; STC-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5160