Learn More
Biomatik Corporation Mouse Proto-Oncogene Wnt-3 (Wnt3) Elisa Kit
Supplier:  Biomatik Corporation EKC37711
Encompass_Preferred
Mouse Proto-Oncogene Wnt-3 (Wnt3) Elisa Kit for detecting Wingless-Type Mmtv Integration Site Family, Member 3 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 39.07-2500pg/ml. Sensitivity: 9.77pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P17553; Wnt3. Target Synonyms: Wnt3; Int-4; Wnt-3; Proto-oncogene Wnt-3; Proto-oncogene Int-4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4464