Learn More
Biomatik Corporation Mouse R-spondin-4(RSPO4) ELISA kit
Supplier:  Biomatik Corporation EKC37756
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 34.4 pg/mL-2200 pg/mL. Sensitivity: 8.6 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q8BJ73. Target Synonyms: RP5-824F16.1, C20orf182, CRISTIN4, FLJ16018, roof plate-specific spondin-4. Restriction: For Research Use Only.
Catalog No. 50-229-4467