Learn More
Biomatik Corporation Mouse serum amyloid A1 (SAA1) ELISA kit
Supplier:  Biomatik Corporation EKC37806
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 3.12 ng/ml - 200 ng/ml. Sensitivity: 0.78 ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P05366. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4469