Learn More
Biomatik Corporation Mouse Signal peptide, CUB and EGF-like domain-containing protein 1(SCUBE1) ELISA kit
Supplier:  Biomatik Corporation EKC37816
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q6NZL8. Target Synonyms: signal peptide-CUB domain-EGF-related 1. Restriction: For Research Use Only.
Catalog No. 50-229-4471