Learn More
Biomatik Corporation Mouse Tumor necrosis factor ligand superfamily member 12(TNFSF12) ELISA kit
Supplier:  Biomatik Corporation EKC37978
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 78 pg/ml - 5000 pg/ml. Sensitivity: 19.5 pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O54907. Target Synonyms: APO3L, DR3LG, MGC129581, MGC20669, TWEAK, APO3/DR3 ligand|TNF-related WEAK inducer of apoptosis. Restriction: For Research Use Only.
Catalog No. 50-229-4480