Learn More
Biomatik Corporation Mouse tumor necrosis factor-like ligand 1 (TL1) ELISA kit
Supplier:  Biomatik Corporation EKC38000
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 62.5 pg/mL-4000 pg/mL. Sensitivity: 15.6 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q5UBV8. Target Synonyms: MGC129934, MGC129935, TL1, TL1A, VEGI, VEGI192A, TNF ligand-related molecule 1|TNF superfamily ligand TL1A|vascular endothelial cell growth inhibitor|vascular endothelial growth inhibitor-192A. Restriction: For Research Use Only.
Catalog No. 50-229-4482