Learn More
Biomatik Corporation Mouse Vascular Endothelial Cell Growth Factor D, Vegf-D Elisa Kit
Supplier:  Biomatik Corporation EKC38028
Encompass_Preferred
Mouse Vascular Endothelial Cell Growth Factor D, Vegf-D Elisa Kit for detecting C-Fos Induced Growth Factor (Vascular Endothelial Growth Factor D) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20pg/ml. Sensitivity: 0.078pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P97946; Vegfd. Target Synonyms: Vegfd; Figf; Vascular endothelial growth factor D; VEGF-D; c-Fos-induced growth factor; FIGF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4484