Learn More
Biomatik Corporation Mouse β-hexosaminidase A,β-Hex A ELISA Kit
Supplier:  Biomatik Corporation EKC38075
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 3.12 μU/mL-200 μU/mL. Sensitivity: 0.78 μU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P29416. Target Synonyms: MGC99608, TSD, N-acetyl-beta-glucosaminidase|beta-N-acetylhexosaminidase|hexosaminidase A|β-hexosaminidase A,β-Hex A|beta-hexosaminidase A,beta-Hex A. Restriction: For Research Use Only.
Catalog No. 50-229-4489