Learn More
Biomatik Corporation Pig complement fragment 5a (C5a) ELISA kit
Supplier:  Biomatik Corporation EKC38148
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Sus scrofa (Pig). Detection Range: 9.375 ng/mL-600 ng/mL. Sensitivity: 2.34 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4499