Learn More
Biomatik Corporation Pig Interleukin 12, Il-12/P70 Elisa Kit
Supplier:  Biomatik Corporation EKC38236
Encompass_Preferred
Pig Interleukin 12, Il-12/P70 Elisa Kit for detecting Interleukin 12, Il-12/P70 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.45-30ng/ml. Sensitivity: 0.11ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q29053; IL12A. Target Synonyms: IL12A; Interleukin-12 subunit alpha; IL-12A; Cytotoxic lymphocyte maturation factor 35 kDa subunit; CLMF p35; IL-12 subunit p35. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4506