Learn More
Biomatik Corporation Rabbit Growth Hormone(GH) ELISA Kit
Supplier:  Biomatik Corporation EKC38446
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 6.25 ng/mL-100 ng/mL. Sensitivity: 6.25 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P46407. Target Synonyms: GH, GH-N, GHN, IGHD1B, hGH-N, pituitary growth hormone. Restriction: For Research Use Only.
Catalog No. 50-229-4523