Learn More
Biomatik Corporation Rabbit Krebs yon den Lundgen-6 (KL-6) ELISA Kit
Supplier:  Biomatik Corporation EKC38481
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 50 IU/mL-800 IU/mL. Sensitivity: 25 IU/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4526