Learn More
Biomatik Corporation Rabbit Parathyroid hormone,PTH ELISA Kit
Supplier:  Biomatik Corporation EKC38512
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.95 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: G1SNZ6. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4532