Learn More
Biomatik Corporation Rabbit Pulmonary surfactant-associated protein B(SFTPB) ELISA kit
Supplier:  Biomatik Corporation EKC38533
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 1.87 ng/mL-120 ng/mL. Sensitivity: 0.46 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P15285. Target Synonyms: PSP-B, SFTB3, SFTP3, SMDP1, SP-B, OTTHUMP00000203253|Pulmonary surfactant-associated protein B, 18kD|surfactant, pulmonary-associated protein B. Restriction: For Research Use Only.
Catalog No. 50-229-4539