Learn More
Biomatik Corporation Rat Arginase 1(ARG1) ELISA Kit
Supplier:  Biomatik Corporation EKC38710
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P07824. Target Synonyms: arginase 1|liver-type arginase|type I arginase. Restriction: For Research Use Only.
Catalog No. 50-229-4554