Learn More
Biomatik Corporation Rat calmodulin,CAM ELISA Kit
Supplier:  Biomatik Corporation EKC38773
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.6 ng/mL-1000 ng/mL. Sensitivity: 3.9 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P0DP29. Target Synonyms: CAMI, DD132, PHKD, calmodulin 1|phosphorylase kinase, delta subunit, CALM1 CALM CAM CAM1, CALM2 CAM2 CAMB, CALM3 CALML2 CAM3 CAMC CAMIII. Restriction: For Research Use Only.
Catalog No. 50-229-4558