Learn More
Biomatik Corporation Rat Fibrinogen Degradation Product,FDP ELISA Kit
Supplier:  Biomatik Corporation EKC39035
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.6 ng/mL-1000 ng/mL. Sensitivity: 3.9 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4576