Learn More
Biomatik Corporation Rat Gamma-glutamyltranspeptidase 1(GGT1) ELISA kit
Supplier:  Biomatik Corporation EKC39082
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 4.7 mIU/mL-300 mIU/mL. Sensitivity: 1.2 mIU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P07314. Target Synonyms: CD224, D22S672, D22S732, GGT, GTG, MGC96892, MGC96904, MGC96963, OTTHUMP00000028921|OTTHUMP00000159078|gamma-glutamyl transpeptidase|glutamyl transpeptidase. Restriction: For Research Use Only.
Catalog No. 50-229-4580