Learn More
Biomatik Corporation Rat Gremlin Elisa Kit
Supplier:  Biomatik Corporation EKC39143
Encompass_Preferred
Rat Gremlin Elisa Kit for detecting Gremlin 1, Cysteine Knot Superfamily, Homolog (Xenopus Laevis) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O35793; Grem1. Target Synonyms: Grem1; Cktsf1b1; Drm; Gremlin-1; Cysteine knot superfamily 1; BMP antagonist 1; Down-regulated in Mos-transformed cells protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4583