Learn More
Biomatik Corporation Rat Hemoglobin (Hb) Elisa Kit
Supplier:  Biomatik Corporation EKC39175
Encompass_Preferred
Rat Hemoglobin (Hb) Elisa Kit for detecting Hemoglobin, Alpha 1 in serum, plasma, lysate for rbc. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ug/ml. Sensitivity: 0.39ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P01946; Hba1. Target Synonyms: Hba1; Hba-a1; Hemoglobin subunit alpha-1/2; Alpha-1/2-globin; Hemoglobin alpha-1/2 chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4590