Learn More
Biomatik Corporation Rat Insulin-Like 3 (Insl3) Elisa Kit
Supplier:  Biomatik Corporation EKC39230
Encompass_Preferred
Rat Insulin-Like 3 (Insl3) Elisa Kit for detecting Insulin-Like 3 (Leydig Cell) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9WUK0; Insl3. Target Synonyms: Insl3; Rlf; Insulin-like 3; Leydig insulin-like peptide; Ley-I-L; Relaxin-like factor) [Cleaved into: Insulin-like 3 B chain; Insulin-like 3 A chain]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4594