Learn More
Biomatik Corporation Rat Interleukin 15, Il-15 Elisa Kit
Supplier:  Biomatik Corporation EKC39257
Encompass_Preferred
Rat Interleukin 15, Il-15 Elisa Kit for detecting Interleukin 15 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P97604; Il15. Target Synonyms: Il15; Interleukin-15; IL-15. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4596