Learn More
Biomatik Corporation Rat keratinocyte autocrine factor/Amphiregulin,KAF/AR ELISA Kit
Supplier:  Biomatik Corporation EKC39312
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 1.25 pg/mL-80 pg/mL. Sensitivity: 0.31 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P24338. Target Synonyms: AR, CRDGF, MGC13647, SDGF, colorectum cell-derived growth factor|schwannoma-derived growth factor. Restriction: For Research Use Only.
Catalog No. 50-229-4598